missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAP3K13 Full-length ORF (NP_004712.1, 1 a.a. - 966 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009175-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16163362

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of serine/threonine protein kinase family. This kinase contains a dual leucine-zipper motif, and has been shown to form dimers/oligomers through its leucine-zipper motif. This kinase can phosphorylate and activate MAPK8/JNK, MAP2K7/MKK7, which suggests a role in the JNK signaling pathway. [provided by RefSeq]

Sequence: MANFQEHLSCSSSPHLPFSESKTFNGLQDELTAMGNHPSPKLLEDQQEKGMVRTELIESVHSPVTTTVLTSVSEDSRDQFENSVLQLREHDESETAVSQGNSNTVDGESTSGTEDIKIQFSRSGSGSGGFLEGLFGCLRPVWNIIGKAYSTDYKLQQQDTWEVPFEEISELQWLGSGAQGAVFLGKFRAEEVAIKKVREQNETDIKHLRKLKHPNIIAFKGVCTQAPCYCIIMEYCAHGQLYEVLRAGRKITPRLLVDWSTGIASGMNYLHLHKIIHRDLKSPNVLVTHTDAVKISDFGTSKELSDKSTKMSFAGTVAWMAPEVIRNEPVSEKVDIWSFGVVLWELLTGEIPYKDVDSSAIIWGVGSNSLHLPVPSTCPDGFKILMKQTWQSKPRNRPSFRQTLMHLDIASADVLATPQETYFKSQAEWREEVKKHFEKIKSEGTCIHRLDEELIRRRREELRHALDIREHYERKLERANNLYMELSAIMLQLEMREKELIKREQAVEKKYPGTYKRHPVRPIIHPNAMEKLMKRKGVPHKSGMQTKRPDLLRSEGIPTTEVAPTASPLSGSPKMSTSSSKSRYRSKPRHRRGNSRGSHSDFAAILKNQPAQENSPHPTYLHQAQSQYPSLHHHNSLQQQYQQPPPAMSQSHHPRLNMHGQDIATCANNLRYFGPAAALRSPLSNHAQRQLPGSSPDLISTAMAADCWRSSEPDKGQAGPWGCCQADAYDPCLQCRPEQYGSLDIPSAEPVGRSPDLSKSPAHNPLLENAQSSEKTEENEFSGCRSESSLGTSHLGTPPALPRKTRPLQKSGDDSSEEEEGEVDSEVEFPRRQRPHRCISSCQSYSTFSSENFSVSDGEEGNTSDHSNSPDELADKLEDRLAEKLDDLLSQTPEIPIDISSHSDGLSDKECAVRRVKTQMSLGKLCVEERGYENPMQFEESDCDSSDGECSDATVRTNKHYSSATW
Specifications

Specifications

NP_004712.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
134.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LZK/MGC133196
MAP3K13
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9175
MAP3K13 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MANFQEHLSCSSSPHLPFSESKTFNGLQDELTAMGNHPSPKLLEDQQEKGMVRTELIESVHSPVTTTVLTSVSEDSRDQFENSVLQLREHDESETAVSQGNSNTVDGESTSGTEDIKIQFSRSGSGSGGFLEGLFGCLRPVWNIIGKAYSTDYKLQQQDTWEVPFEEISELQWLGSGAQGAVFLGKFRAEEVAIKKVREQNETDIKHLRKLKHPNIIAFKGVCTQAPCYCIIMEYCAHGQLYEVLRAGRKITPRLLVDWSTGIASGMNYLHLHKIIHRDLKSPNVLVTHTDAVKISDFGTSKELSDKSTKMSFAGTVAWMAPEVIRNEPVSEKVDIWSFGVVLWELLTGEIPYKDVDSSAIIWGVGSNSLHLPVPSTCPDGFKILMKQTWQSKPRNRPSFRQTLMHLDIASADVLATPQETYFKSQAEWREEVKKHFEKIKSEGTCIHRLDEELIRRRREELRHALDIREHYERKLERANNLYMELSAIMLQLEMREKELIKREQAVEKKYPGTYKRHPVRPIIHPNAMEKLMKRKGVPHKSGMQTKRPDLLRSEGIPTTEVAPTASPLSGSPKMSTSSSKSRYRSKPRHRRGNSRGSHSDFAAILKNQPAQENSPHPTYLHQAQSQYPSLHHHNSLQQQYQQPPPAMSQSHHPRLNMHGQDIATCANNLRYFGPAAALRSPLSNHAQRQLPGSSPDLISTAMAADCWRSSEPDKGQAGPWGCCQADAYDPCLQCRPEQYGSLDIPSAEPVGRSPDLSKSPAHNPLLENAQSSEKTEENEFSGCRSESSLGTSHLGTPPALPRKTRPLQKSGDDSSEEEEGEVDSEVEFPRRQRPHRCISSCQSYSTFSSENFSVSDGEEGNTSDHSNSPDELADKLEDRLAEKLDDLLSQTPEIPIDISSHSDGLSDKECAVRRVKTQMSLGKLCVEERGYENPMQFEESDCDSSDGECSDATVRTNKHYSSATW
RUO
MAP3K13
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers