missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human High Sensitivity Cytokine Panel B Luminex™ Performance Control
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Ea.
Beschreibung
- For use as quantitative controls for the determination of human cytokine concentrations in biological fluids.
- Concentrations have been assigned using R&D Systems Luminex™ High Performance Assays.
- Controls are prepared in a buffered protein solution with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
Spezifikation
Spezifikation
| Product Type | ELISA Kit |
| Quantity | 1 Ea. |
| Species | Human |
| Binding Target | Luminex Assay Controls |
| For Use With (Equipment) | Human Magnetic HS Cytokine B Panel |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur