missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAGED2 Partial ORF (NP_055414, 16 a.a. - 125 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010916-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193366

  • 528.00€ / 25µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

This gene is a member of the MAGED gene family. While the MAGEA and MAGEB genes are silent in normal tissues with the exception of testis and placenta, the MAGED genes are expressed ubiquitously. The MAGED genes are clustered on chromosome Xp11. This gene is located in Xp11.2, a hot spot for X-linked mental retardation (XLMR). Multiple alternatively spliced transcript variants have been found for this gene, however, the full length nature of some variants has not been defined. [provided by RefSeq]

Sequence: AEASEKDSSSMMQTLLTVTQNVEVPETPKASKALEVSEDVKVSKASGVSKATEVSKTPEAREAPATQASSTTQLTDTQVLAAENKSLAADTKKQNADPQAVTMPATETKK
Specifications

Specifications

NP_055414
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AEASEKDSSSMMQTLLTVTQNVEVPETPKASKALEVSEDVKVSKASGVSKATEVSKTPEAREAPATQASSTTQLTDTQVLAAENKSLAADTKKQNADPQAVTMPATETKK
RUO
MAGED2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10916
MAGED2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
11B6/BCG1/HCA10/JCL-1/MAGE-D2/MAGED/MGC8386
MAGED2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers