Learn More
Abnova™ Human MAGEB4 Full-length ORF (AAH32852.1, 1 a.a. - 346 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004115-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEB genes are clustered on chromosome Xp22-p21. This gene sequence ends in the first intron of MAGEB1, another family member. This gene is expressed in testis. [provided by RefSeq]
Sequence: MPRGQKSKLRAREKRQRTRGQTQDLKVGQPTAAEKEESPSPSSSVLRDTASSSLAFGIPQEPQREPPTTSAAAAMSCTGSDKGDESQDEENASSSQASTSTERSLKDSLTRKTKMLVQFLLYKYKMKEPTTKAEMLKIISKKYKEHFPEIFRKVSQRTELVFGLALKEVNPTTHSYILVSMLGPNYGNQSSAWTLPRNGLLMPLLSVIFLNGNCAREEEIWEFLNMLGIYDGKRHLIFGEPRKLITQDLVQEKYLEYQQVPNSDPPRYQFLWGPRAHAETSKMKVLEFLAKVNDTTPNNFPLLYEEALRDEEERAGARPRVAARRGTTAMTSAYSRATSSSSSQPMSpecifications
AAH32852.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC33144 | |
MAGEB4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4115 | |
MAGEB4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPRGQKSKLRAREKRQRTRGQTQDLKVGQPTAAEKEESPSPSSSVLRDTASSSLAFGIPQEPQREPPTTSAAAAMSCTGSDKGDESQDEENASSSQASTSTERSLKDSLTRKTKMLVQFLLYKYKMKEPTTKAEMLKIISKKYKEHFPEIFRKVSQRTELVFGLALKEVNPTTHSYILVSMLGPNYGNQSSAWTLPRNGLLMPLLSVIFLNGNCAREEEIWEFLNMLGIYDGKRHLIFGEPRKLITQDLVQEKYLEYQQVPNSDPPRYQFLWGPRAHAETSKMKVLEFLAKVNDTTPNNFPLLYEEALRDEEERAGARPRVAARRGTTAMTSAYSRATSSSSSQPM | |
RUO | |
MAGEB4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |