missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LRRN3 Full-length ORF (NP_060804.2, 1 a.a. - 708 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054674-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100693

  • 508.00€ / 25µg

Estimated Shipment: 12-05-2023
to see stock.
Add to basket

Description

Description

Sequence: MKDMPLRIHVLLGLAITTLVQAVDKKVDCPRLCTCEIRPWFTPRSIYMEASTVDCNDLGLLTFPARLPANTQILLLQTNNIAKIEYSTDFPVNLTSLDLSQNNLSSVTNINVKKMPQLLSVYLEENKLTELPEKCLSELSNLQELYINHNLLSTISPGAFIGLHNLLRLHLNSNRLQMINSKWFDALPNLEILMIGENPIIRIKDMNFKPLINLRSLVIAGINLTEIPDNALVGLENLESISFYDNRLIKVPHVALQKVVNLKFLDLNKNPINRIRRGDFSNMLHLKELGINNMPELISIDSLAVDNLPDLRKIEATNNPRLSYIHPNAFFRLPKLESLMLNSNALSALYHGTIESLPNLKEISIHSNPIRCDCVIRWMNMNKTNIRFMEPDSLFCVDPPEFQGQNVRQVHFRDMMEICLPLIAPESFPSNLNVEAGSYVSFHCRATAEPQPEIYWITPSGQKLLPNTLTDKFYVHSEGTLDINGVTPKEGGLYTCIATNLVGADLKSVMIKVDGSFPQDNNGSLNIKIRDIHANSVLVSWKASSKILKSSVKWTAFVKTENSHAAQSARIPSDVKVYNLTHLNPSTEYKICIDIPTIYQKNRKKCVNVTTKGLHPDQKEYEKNNTTTLMACLGGLLGIIGVICLISCLSPEMNCDGGHSYVRNYLQKPTFALGELYPPLINLWEAGKEKSTSLKVKATVIGLPTNMS
Specifications

Specifications

NP_060804.2
Liquid
54674
LRRN3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKDMPLRIHVLLGLAITTLVQAVDKKVDCPRLCTCEIRPWFTPRSIYMEASTVDCNDLGLLTFPARLPANTQILLLQTNNIAKIEYSTDFPVNLTSLDLSQNNLSSVTNINVKKMPQLLSVYLEENKLTELPEKCLSELSNLQELYINHNLLSTISPGAFIGLHNLLRLHLNSNRLQMINSKWFDALPNLEILMIGENPIIRIKDMNFKPLINLRSLVIAGINLTEIPDNALVGLENLESISFYDNRLIKVPHVALQKVVNLKFLDLNKNPINRIRRGDFSNMLHLKELGINNMPELISIDSLAVDNLPDLRKIEATNNPRLSYIHPNAFFRLPKLESLMLNSNALSALYHGTIESLPNLKEISIHSNPIRCDCVIRWMNMNKTNIRFMEPDSLFCVDPPEFQGQNVRQVHFRDMMEICLPLIAPESFPSNLNVEAGSYVSFHCRATAEPQPEIYWITPSGQKLLPNTLTDKFYVHSEGTLDINGVTPKEGGLYTCIATNLVGADLKSVMIKVDGSFPQDNNGSLNIKIRDIHANSVLVSWKASSKILKSSVKWTAFVKTENSHAAQSARIPSDVKVYNLTHLNPSTEYKICIDIPTIYQKNRKKCVNVTTKGLHPDQKEYEKNNTTTLMACLGGLLGIIGVICLISCLSPEMNCDGGHSYVRNYLQKPTFALGELYPPLINLWEAGKEKSTSLKVKATVIGLPTNMS
RUO
LRRN3
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
105.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FIGLER5/FLJ11129/NLRR-3/NLRR3
LRRN3
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers