missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LRRC40 Full-length ORF (NP_060238.3, 1 a.a. - 602 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055631-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101623

  • 335.00€ / 10µg

Estimated Shipment: 12-05-2023
to see stock.
Add to basket

Description

Description

Sequence: MSRLKRIAGQDLRAGFKAGGRDCGTSVPQGLLKAARKSGQLNLSGRNLSEVPQCVWRINVDIPEEANQNLSFGATERWWEQTDLTKLIISNNKLQSLTDDLRLLPALTVLDIHDNQLTSLPSAIRELENLQKLNVSHNKLKILPEEITNLRNLKCLYLQHNELTCISEGFEQLSNLEDLDLSNNHLTTVPASFSSLSSLVRLNLSSNELKSLPAEINRMKRLKHLDCNSNLLETIPPELAGMESLELLYLRRNKLRFLPEFPSCSLLKELHVGENQIEMLEAEHLKHLNSILVLDLRDNKLKSVPDEIILLRSLERLDLSNNDISSLPYSLGNLHLKFLALEGNPLRTIRREIISKGTQEVLKYLRSKIKDDGPSQSESATETAMTLPSESRVNIHAIITLKILDYSDKQATLIPDEVFDAVKSNIVTSINFSKNQLCEIPKRMVELKEMVSDVDLSFNKLSFISLELCVLQKLTFLDLRNNFLNSLPEEMESLVRLQTINLSFNRFKMLPEVLYRIFTLETILISNNQVGSVDPQKMKMMENLTTLDLQNNDLLQIPPELGNCVNLRTLLLDGNPFRVPRAAILMKGTAAILEYLRDRIPT
Specifications

Specifications

NP_060238.3
Liquid
55631
LRRC40 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSRLKRIAGQDLRAGFKAGGRDCGTSVPQGLLKAARKSGQLNLSGRNLSEVPQCVWRINVDIPEEANQNLSFGATERWWEQTDLTKLIISNNKLQSLTDDLRLLPALTVLDIHDNQLTSLPSAIRELENLQKLNVSHNKLKILPEEITNLRNLKCLYLQHNELTCISEGFEQLSNLEDLDLSNNHLTTVPASFSSLSSLVRLNLSSNELKSLPAEINRMKRLKHLDCNSNLLETIPPELAGMESLELLYLRRNKLRFLPEFPSCSLLKELHVGENQIEMLEAEHLKHLNSILVLDLRDNKLKSVPDEIILLRSLERLDLSNNDISSLPYSLGNLHLKFLALEGNPLRTIRREIISKGTQEVLKYLRSKIKDDGPSQSESATETAMTLPSESRVNIHAIITLKILDYSDKQATLIPDEVFDAVKSNIVTSINFSKNQLCEIPKRMVELKEMVSDVDLSFNKLSFISLELCVLQKLTFLDLRNNFLNSLPEEMESLVRLQTINLSFNRFKMLPEVLYRIFTLETILISNNQVGSVDPQKMKMMENLTTLDLQNNDLLQIPPELGNCVNLRTLLLDGNPFRVPRAAILMKGTAAILEYLRDRIPT
RUO
LRRC40
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
94.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20331/RP4-677H15.1/dJ677H15.1
LRRC40
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers