missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LOC150577 Full-length ORF (XP_379114.1, 1 a.a. - 92 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00150577-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16158623

  • 335.00€ / 10µg
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MVHSRKWGESGQRCPGGLCLHHLERHRPFSLHSLFRKEAYSQMNAPRLRDKANPWDAEGTWITSARERSPNSASRRPEVWPPVDTQLRDEQQ
Specifications

Specifications

XP_379114.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LOC150577
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
150577
LOC150577 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVHSRKWGESGQRCPGGLCLHHLERHRPFSLHSLFRKEAYSQMNAPRLRDKANPWDAEGTWITSARERSPNSASRRPEVWPPVDTQLRDEQQ
RUO
LOC150577
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers