Learn More
Abnova™ Human LMX1A Full-length ORF (NP_001028679.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004009-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Insulin is produced exclusively by the beta cells in the islets of Langerhans in the pancreas. The level and beta-cell specificity of insulin gene expression are regulated by a set of nuclear genes that bind to specific sequences within the promoter of the insulin gene (INS; MIM 176730) and interact with RNA polymerase to activate or repress transcription. LMX1 is a homeodomain protein that binds an A/T-rich sequence in the insulin promoter and stimulates transcription of insulin (German et al., 1994 [PubMed 7698771]).[supplied by OMIM]
Sequence: MEGIMNPYTALPTPQQLLAIEQSVYSSDPFRQGLTPPQMPGDHMHPYGAEPLFHDLDSDDTSLSNLGDCFLATSEAGPLQSRVGNPIDHLYSMQNSYFTSSpecifications
NP_001028679.1 | |
Liquid | |
4009 | |
LMX1A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEGIMNPYTALPTPQQLLAIEQSVYSSDPFRQGLTPPQMPGDHMHPYGAEPLFHDLDSDDTSLSNLGDCFLATSEAGPLQSRVGNPIDHLYSMQNSYFTS | |
RUO | |
LMX1A | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LMX-1/LMX1/LMX1.1/MGC87616 | |
LMX1A | |
Yes | |
wheat germ expression system |