Learn More
Abnova™ Human LMBR1 Full-length ORF (NP_071903.2, 1 a.a. - 490 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064327-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the LMBR1-like membrane protein family. Another member of this protein family has been shown to be a lipocalin transmembrane receptor. A highly conserved, cis-acting regulatory module for the sonic hedgehog gene is located within an intron of this gene. Consequently, disruption of this genic region can alter sonic hedgehog expression and affect limb patterning, but it is not known if this gene functions directly in limb development. Mutations and chromosomal deletions and rearrangements in this genic region are associated with acheiropody and preaxial polydactyly, which likely result from altered sonic hedgehog expression. [provided by RefSeq]
Sequence: MEGQDEVSAREQHFHSQVRESTICFLLFAILYVVSYFIITRYKRKSDEQEDEDAIVNRISLFLSTFTLAVSAGAVLLLPFSIISNEILLSFPQNYYIQWLNGSLIHGLWNLASLFSNLCLFVLMPFAFFFLESEGFAGLKKGIRARILETLVMLLLLALLILGIVWVASALIDNDAASMESLYDLWEFYLPYLYSCISLMGCLLLLLCTPVGLSRMFTVMGQLLVKPTILEDLDEQIYIITLEEEALQRRLNGLSSSVEYNIMELEQELENVKTLKTKLERRKKASAWERNLVYPAVMVLLLIETSISVLLVACNILCLLVDETAMPKGTRGPGIGNASLSTFGFVGAALEIILIFYLMVSSVVGFYSLRFFGNFTPKKDDTTMTKIIGNCVSILVLSSALPVMSRTLGITRFDLLGDFGRFNWLGNFYIVLSYNLLFAIVTTLCLVRKFTSAVREELFKALGLHKLHLPNTSRDSETAKPSVNGHQKALSpecifications
NP_071903.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACHP/C7orf2/DIF14/FLJ11665/PPD2/TPT | |
LMBR1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64327 | |
LMBR1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEGQDEVSAREQHFHSQVRESTICFLLFAILYVVSYFIITRYKRKSDEQEDEDAIVNRISLFLSTFTLAVSAGAVLLLPFSIISNEILLSFPQNYYIQWLNGSLIHGLWNLASLFSNLCLFVLMPFAFFFLESEGFAGLKKGIRARILETLVMLLLLALLILGIVWVASALIDNDAASMESLYDLWEFYLPYLYSCISLMGCLLLLLCTPVGLSRMFTVMGQLLVKPTILEDLDEQIYIITLEEEALQRRLNGLSSSVEYNIMELEQELENVKTLKTKLERRKKASAWERNLVYPAVMVLLLIETSISVLLVACNILCLLVDETAMPKGTRGPGIGNASLSTFGFVGAALEIILIFYLMVSSVVGFYSLRFFGNFTPKKDDTTMTKIIGNCVSILVLSSALPVMSRTLGITRFDLLGDFGRFNWLGNFYIVLSYNLLFAIVTTLCLVRKFTSAVREELFKALGLHKLHLPNTSRDSETAKPSVNGHQKAL | |
RUO | |
LMBR1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |