missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LMAN2 Full-length ORF (NP_006807.1, 1 a.a. - 356 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010960-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195922

  • 528.00€ / 25µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a type I transmembrane lectin that shuttles between the endoplasmic reticulum, the Golgi apparatus and the plasma membrane. The encoded protein binds high mannose type glycoproteins and may facilitate their sorting, trafficking and quality control. [provided by RefSeq]

Sequence: MAAEGWIWRWGWGRRCLGRPGLLGPGPGPTTPLFLLLLLGSVTADITDGNSEHLKREHSLIKPYQGVGSSSMPLWDFQGSTMLTSQYVRLTPDERSKEGSIWNHQPCFLKDWEMHVHFKVHGTGKKNLHGDGIALWYTRDRLVPGPVFGSKDNFHGLAIFLDTYPNDETTERVFPYISVMVNNGSLSYDHSKDGRWTELAGCTADFRNRDHDTFLAVRYSRGRLTVMTDLEDKNEWKNCIDITGVRLPTGYYFGASAGTGDLSDNHDIISMKLFQLMVEHTPDEESIDWTKIEPSVNFLKSPKDNVDDPTGNFRSGPLTGWRVFLLLLCALLGIVVCAVVGAVVFQKRQERNKRFY
Specifications

Specifications

NP_006807.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C5orf8/GP36B/VIP36
LMAN2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10960
LMAN2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAEGWIWRWGWGRRCLGRPGLLGPGPGPTTPLFLLLLLGSVTADITDGNSEHLKREHSLIKPYQGVGSSSMPLWDFQGSTMLTSQYVRLTPDERSKEGSIWNHQPCFLKDWEMHVHFKVHGTGKKNLHGDGIALWYTRDRLVPGPVFGSKDNFHGLAIFLDTYPNDETTERVFPYISVMVNNGSLSYDHSKDGRWTELAGCTADFRNRDHDTFLAVRYSRGRLTVMTDLEDKNEWKNCIDITGVRLPTGYYFGASAGTGDLSDNHDIISMKLFQLMVEHTPDEESIDWTKIEPSVNFLKSPKDNVDDPTGNFRSGPLTGWRVFLLLLCALLGIVVCAVVGAVVFQKRQERNKRFY
RUO
LMAN2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers