Learn More
Abnova™ Human LIFR Partial ORF (NP_002301, 45 a.a. - 154 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003977-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that belongs to the type I cytokine receptor family. This protein combines with a high-affinity converter subunit, gp130, to form a receptor complex that mediates the action of the leukemia inhibitory factor, a polyfunctional cytokine that is involved in cellular differentiation, proliferation and survival in the adult and the embryo. Mutations in this gene cause Schwartz-Jampel syndrome type 2, a disease belonging to the group of the bent-bone dysplasias. A translocation that involves the promoter of this gene, t(5;8)(p13;q12) with the pleiomorphic adenoma gene 1, is associated with salivary gland pleiomorphic adenoma, a common type of benign epithelial tumor of the salivary gland. Multiple splice variants encoding the same protein have been found for this gene. [provided by RefSeq]
Sequence: QKKGAPHDLKCVTNNLQVWNCSWKAPSGTGRGTDYEVCIENRSRSCYQLEKTSIKIPALSHGDYEITINSLHDFGSSTSKFTLNEQNVSLIPDTPEILNLSADFSTSTLYSpecifications
NP_002301 | |
Liquid | |
3977 | |
LIFR (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD118/FLJ98106/FLJ99923/LIF-R/SJS2/STWS/SWS | |
LIFR | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QKKGAPHDLKCVTNNLQVWNCSWKAPSGTGRGTDYEVCIENRSRSCYQLEKTSIKIPALSHGDYEITINSLHDFGSSTSKFTLNEQNVSLIPDTPEILNLSADFSTSTLY | |
RUO | |
LIFR | |
Wheat Germ (in vitro) | |
GST |