Learn More
Abcam Human LGALS9C knockout A549 cell line
Product Code. 32141341
Change view
Click to view available options
2x1000000 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
32141341 2x1000000 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32141341 Supplier Abcam Supplier No. ab2671892x1000000Cells_
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
LGALS9C KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 22 bp deletion in exon 3 and 4 bp deletion in exon 3 and 4 bp insertion in exon 3 and 8 bp deletion in exon 3. To order both knockout and wild-type control ce...
LGALS9C KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 22 bp deletion in exon 3 and 4 bp deletion in exon 3 and 4 bp insertion in exon 3 and 8 bp deletion in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells