missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LGALS14 Full-length ORF (NP_982297.1, 1 a.a. - 168 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00056891-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192293

  • 349.00€ / 10µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

This gene is predominantly expressed in placenta. The encoded protein belongs to the galectin (galaptin/S-lectin) family. The members of galectin family contain one or two carbohydrate recognition domains, which can bind beta-galactoside. Two alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]

Sequence: MSLTHRLHLCKYWGCAVSNVCRFWEGRPLPLMIVVPYTLPVSLPVGSCVIITGTPILTFVKDPQLEVNFYTGMDEDSDIAFQFRLHFGHPAIMNSCVFGIWRYEEKCYYLPFEDGKPFELCIYVRHKEYKVMVNGQRIYNFAHRFPPASVKMLQVFRDISLTRVLISD
Specifications

Specifications

NP_982297.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
46kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CLC2/MGC22235/PPL13
LGALS14
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
56891
LGALS14 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSLTHRLHLCKYWGCAVSNVCRFWEGRPLPLMIVVPYTLPVSLPVGSCVIITGTPILTFVKDPQLEVNFYTGMDEDSDIAFQFRLHFGHPAIMNSCVFGIWRYEEKCYYLPFEDGKPFELCIYVRHKEYKVMVNGQRIYNFAHRFPPASVKMLQVFRDISLTRVLISD
RUO
LGALS14
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers