missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human LDL
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
- This product is a human plasma-derived low-density lipoprotein (LDL)-based culture supplement that has been pre-screened for the culture, expansion, and colony assay of human hematopoietic and non-hematopoietic cells in serum-free culture media
- It promotes the proliferation and survival of human hematopoietic and other progenitor cells in culture, resulting in increased cell output in expansion cultures and increased colony numbers and/or size in colony assays
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture |
| Quantity | 5 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction