Learn More
Abnova™ Human LDHC Partial ORF (NP_002292.1, 71 a.a. - 160 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003948-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Lactate dehydrogenase C catalyzes the conversion of L-lactate and NAD to pyruvate and NADH in the final step of anaerobic glycolysis. LDHC is testis-specific and belongs to the lactate dehydrogenase family. Two transcript variants have been detected which differ in the 5' untranslated region. [provided by RefSeq]
Sequence: FFSTSKITSGKDYSVSANSRIVIVTAGARQQEGETRLALVQRNVAIMKSIIPAIVHYSPDCKILVVSNPVDILTYIVWKISGLPVTRVIGSpecifications
NP_002292.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FFSTSKITSGKDYSVSANSRIVIVTAGARQQEGETRLALVQRNVAIMKSIIPAIVHYSPDCKILVVSNPVDILTYIVWKISGLPVTRVIG | |
RUO | |
LDHC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3948 | |
LDHC (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CT32/LDH3/LDHX/MGC111073 | |
LDHC | |
Recombinant | |
wheat germ expression system |