missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LCOR Full-length ORF (AAH53359.1, 1 a.a. - 406 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084458-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195903

  • 349.00€ / 10µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

LCOR is a transcriptional corepressor widely expressed in fetal and adult tissues that is recruited to agonist-bound nuclear receptors through a single LxxLL motif, also referred to as a nuclear receptor (NR) box (Fernandes et al., 2003 [PubMed 12535528]).[supplied by OMIM]

Sequence: MQRMIQQFAAEYTSKNSSTQDPSQPNSTKNQSLPKASPVTTSPTAATTQNPVLSKLLMADQDSPLDLTVRKSQSEPSEQDGVLDLSTKKSPCAGSTSLSHSPGCSSTQGNGRPGRPSQYRPDGLRSGDGVPPRSLQDGTREGFGHSTSLKVPLARSLQISEELLSRNQLSTAASLGPSGLQNHGQHLILSREASWAKPHYEFNLSRMKFRGNGALSNISDLPFLAENSAFPKMALQAKQDGKKDVSHSSPVDLKIPQVRGMDLSWESRTGDQYSYSSLVMGSQTESALSKKLRAILPKQSRKSMLDAGPDSWGSDAEQSTSGQPYPTSDQEGDPGSKQPRKKRGRYRQYNSEILEEAISVVMSGKMSVSKAQSIYGIPHSTLEYKVKERLGTLKNPPKKKMKLMSG
Specifications

Specifications

AAH53359.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ38026/KIAA1795/MLR2/RP11-175O19.1
LCOR
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84458
LCOR (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQRMIQQFAAEYTSKNSSTQDPSQPNSTKNQSLPKASPVTTSPTAATTQNPVLSKLLMADQDSPLDLTVRKSQSEPSEQDGVLDLSTKKSPCAGSTSLSHSPGCSSTQGNGRPGRPSQYRPDGLRSGDGVPPRSLQDGTREGFGHSTSLKVPLARSLQISEELLSRNQLSTAASLGPSGLQNHGQHLILSREASWAKPHYEFNLSRMKFRGNGALSNISDLPFLAENSAFPKMALQAKQDGKKDVSHSSPVDLKIPQVRGMDLSWESRTGDQYSYSSLVMGSQTESALSKKLRAILPKQSRKSMLDAGPDSWGSDAEQSTSGQPYPTSDQEGDPGSKQPRKKRGRYRQYNSEILEEAISVVMSGKMSVSKAQSIYGIPHSTLEYKVKERLGTLKNPPKKKMKLMSG
RUO
LCOR
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers