Learn More
Abnova™ Human LCOR Full-length ORF (AAH53359.1, 1 a.a. - 406 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084458-P01.25ug
Additional Details : Weight : 0.00010kg
Description
LCOR is a transcriptional corepressor widely expressed in fetal and adult tissues that is recruited to agonist-bound nuclear receptors through a single LxxLL motif, also referred to as a nuclear receptor (NR) box (Fernandes et al., 2003 [PubMed 12535528]).[supplied by OMIM]
Sequence: MQRMIQQFAAEYTSKNSSTQDPSQPNSTKNQSLPKASPVTTSPTAATTQNPVLSKLLMADQDSPLDLTVRKSQSEPSEQDGVLDLSTKKSPCAGSTSLSHSPGCSSTQGNGRPGRPSQYRPDGLRSGDGVPPRSLQDGTREGFGHSTSLKVPLARSLQISEELLSRNQLSTAASLGPSGLQNHGQHLILSREASWAKPHYEFNLSRMKFRGNGALSNISDLPFLAENSAFPKMALQAKQDGKKDVSHSSPVDLKIPQVRGMDLSWESRTGDQYSYSSLVMGSQTESALSKKLRAILPKQSRKSMLDAGPDSWGSDAEQSTSGQPYPTSDQEGDPGSKQPRKKRGRYRQYNSEILEEAISVVMSGKMSVSKAQSIYGIPHSTLEYKVKERLGTLKNPPKKKMKLMSGSpecifications
AAH53359.1 | |
Liquid | |
84458 | |
LCOR (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQRMIQQFAAEYTSKNSSTQDPSQPNSTKNQSLPKASPVTTSPTAATTQNPVLSKLLMADQDSPLDLTVRKSQSEPSEQDGVLDLSTKKSPCAGSTSLSHSPGCSSTQGNGRPGRPSQYRPDGLRSGDGVPPRSLQDGTREGFGHSTSLKVPLARSLQISEELLSRNQLSTAASLGPSGLQNHGQHLILSREASWAKPHYEFNLSRMKFRGNGALSNISDLPFLAENSAFPKMALQAKQDGKKDVSHSSPVDLKIPQVRGMDLSWESRTGDQYSYSSLVMGSQTESALSKKLRAILPKQSRKSMLDAGPDSWGSDAEQSTSGQPYPTSDQEGDPGSKQPRKKRGRYRQYNSEILEEAISVVMSGKMSVSKAQSIYGIPHSTLEYKVKERLGTLKNPPKKKMKLMSG | |
RUO | |
LCOR | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ38026/KIAA1795/MLR2/RP11-175O19.1 | |
LCOR | |
Yes | |
wheat germ expression system |