missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LCN1 Partial ORF (NP_002288, 24 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003933-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100605

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene belongs to the lipocalin family. Lipocalins are a group of extracellular proteins that are able to bind lipophiles by enclosure within their structures to minimize solvent contact. This protein may bind hydrophobic ligands and inhibit cysteine proteinases. It may also play a role in taste reception. [provided by RefSeq]

Sequence: SDEEIQDVSGTWYLKAMTVDREFPEMNLESVTPMTLTTLEGGNLEAKVTMLISGRCQEVKAVLEKTDEPGKYTADGGKHVAYIIRSHVKDHYIFYCE
Specifications

Specifications

NP_002288
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SDEEIQDVSGTWYLKAMTVDREFPEMNLESVTPMTLTTLEGGNLEAKVTMLISGRCQEVKAVLEKTDEPGKYTADGGKHVAYIIRSHVKDHYIFYCE
RUO
LCN1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3933
LCN1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC71975/PMFA/TP/VEGP
LCN1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers