Learn More
Abnova™ Human LARS2 Partial ORF (NP_056155, 806 a.a. - 903 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023395-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a class 1 aminoacyl-tRNA synthetase, mitochondrial leucyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. [provided by RefSeq]
Sequence: VPRKLCAHYTWDASVLLQAWPAVDPEFLQQPEVVQMAVLINNKACGKIPVPQQVARDQDKVHEFVLQSELGVRLLQGRSIKKSFLSPRTALINFLVQDSpecifications
NP_056155 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VPRKLCAHYTWDASVLLQAWPAVDPEFLQQPEVVQMAVLINNKACGKIPVPQQVARDQDKVHEFVLQSELGVRLLQGRSIKKSFLSPRTALINFLVQD | |
RUO | |
LARS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23395 | |
LARS2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0028/LEURS/MGC26121 | |
LARS2 | |
Recombinant | |
wheat germ expression system |