missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Lamin B1 ELISA Kit (Colorimetric)

Product Code. 30239662 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30239662 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30239662 Supplier Novus Biologicals™ Supplier No. NBP339744

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Colorimetric ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.312 - 20 ng/mL (example only; lot dependent)
Assay Sensitivity 0.118 ng/mL (example only; lot dependent)
Detection Method Colorimetric
Immunoassay Kit Format Pre-coated 96T Strip Plate
Product Type ELISA Kit (Colorimetric)
Sample Type Tissue Homogenates, Cell Lysates, Other Biological Fluids
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 4001
Gene Symbol LMNB1
Quantity 1 Kit
Research Discipline Apoptosis, Cancer, Core ESC Like Genes, Loading Controls, Phospho Specific, Stem Cell Markers, Tumor Suppressors
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, MGC111419
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.