Learn More
ACROBiosystems Human LAG-3 / CD223 Protein, His Tag, low endotoxin
low endotoxin
Brand: ACROBiosystems LA3-H522A-1MG
Additional Details : Weight : 0.00001kg
Description
Lymphocyte activation gene 3 protein (LAG3) is also known as CD antigen CD223 and protein FDC, which belongs to immunoglobulin (Ig) superfamily and contains 4 extracellular Ig-like domains. The LAG3 gene contains 8 exons. The sequence data, exon/intron organization, and chromosomal localization all indicate a close relationship of LAG3 to CD4. LAG3 /CD223 involved in lymphocyte activation. LAG3 /CD223 binds to HLA class-II antigens.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.1 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_002277.4 | |
1 mg | |
Leu 23 - Leu 450 | |
LAG3,CD223,FDC | |
Unconjugated | |
His Tag | |
Less than 0.01 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
47.1 kDa | |
Human LAG-3, His Tag, low endotoxin (LA3-H522a) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession # P18627-1). | |
-20°C | |
LAG-3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |