Learn More
Abnova™ Human KRT35 Full-length ORF (AAI46509.1, 1 a.a. - 455 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003886-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the keratin gene family. This type I hair keratin is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. [provided by RefSeq]
Sequence: MASKCLKAGFSSGSLKSPGGASGGSTRVSAMYSSSSCKLPSLSPVARSFSACSVGLGRSSYRATSCLPALCLPAGGFATSYSGGGGWFGEGILTGNEKETMQSLNDRLAGYLEKVRQLEQENASLESRIREWCEQQVPYMCPDYQSYFRTIEELQKKTLCSKAENARLVVEIDNAKLAADDFRTKYETEVSLRQLVESDINGLRRILDDLTLCKSDLEAQVESLKEELLCLKKNHEEEVNSLRCQLGDRLNVEVDAAPPVDLNRVLEEMRCQYETLVENNRRDAEDWLDTQSEELNQQVVSSSEQLQSCQAEIIELRRTVNALEIELQAQHSMRDALESTLAETEARYSSQLAQMQCMITNVEAQLAEIRADLERQNQEYQVLLDVRARLECEINTYRGLLESEDSKLPCNPCAPDYSPSKSCLPCLPAASCGPSAARTNCSPRPICVPCPGGRFSpecifications
AAI46509.1 | |
Liquid | |
3886 | |
KRT35 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASKCLKAGFSSGSLKSPGGASGGSTRVSAMYSSSSCKLPSLSPVARSFSACSVGLGRSSYRATSCLPALCLPAGGFATSYSGGGGWFGEGILTGNEKETMQSLNDRLAGYLEKVRQLEQENASLESRIREWCEQQVPYMCPDYQSYFRTIEELQKKTLCSKAENARLVVEIDNAKLAADDFRTKYETEVSLRQLVESDINGLRRILDDLTLCKSDLEAQVESLKEELLCLKKNHEEEVNSLRCQLGDRLNVEVDAAPPVDLNRVLEEMRCQYETLVENNRRDAEDWLDTQSEELNQQVVSSSEQLQSCQAEIIELRRTVNALEIELQAQHSMRDALESTLAETEARYSSQLAQMQCMITNVEAQLAEIRADLERQNQEYQVLLDVRARLECEINTYRGLLESEDSKLPCNPCAPDYSPSKSCLPCLPAASCGPSAARTNCSPRPICVPCPGGRF | |
RUO | |
KRT35 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HA5/Ha-5/KRTHA5/hHa5 | |
KRT35 | |
Yes | |
wheat germ expression system |