missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KRT28 Full-length ORF (AAI48795.1, 1 a.a. - 464 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00162605-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16158943

  • 335.00€ / 10µg
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the type I (acidic) keratin family, which belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. The type I keratin genes are clustered in a region of chromosome 17q12-q21. [provided by RefSeq]

Sequence: MSLQFSNGSRHVCLRSGAGSVRPLNGGAGFAGSSACGGSVAGSEFSCALGGGLGSVPGGSHAGGALGNAACIGFAGSEGGLLSGNEKVTMQNLNDRLASYLDNVRALEEANAELERKIKGWYEKYGPGSCRGLDHDYSRYHLTIEDLKNKIISSTTTNANVILQIDNARLAADDFRLKYENELTLHQNVEADINGLRRVLDELTLCRTDQELQYESLSEEMTYLKKNHEEEMKALQCAAGGNVNVEMNAAPGVDLAVLLNNMRAEYEALAEQNRKDAEAWFNEKSASLQQQISHDSGAATFARSQLTEMRRTLQTLEIQLQSLMATKHSLECSLTETESNYCTQLAQIQAQIGALEEQLHQVRTETEGQKLEYEHLLDVKVHLEKEIETYCRLIDGDGNSCSKSKGFGSGSPGNSSKDLSKTTLVKTVVEELDQRGKVLSSRIHSIEEKTSKMTNGKTEQRVPF
Specifications

Specifications

AAI48795.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
77.99kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KRT25D
KRT28
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
162605
KRT28 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSLQFSNGSRHVCLRSGAGSVRPLNGGAGFAGSSACGGSVAGSEFSCALGGGLGSVPGGSHAGGALGNAACIGFAGSEGGLLSGNEKVTMQNLNDRLASYLDNVRALEEANAELERKIKGWYEKYGPGSCRGLDHDYSRYHLTIEDLKNKIISSTTTNANVILQIDNARLAADDFRLKYENELTLHQNVEADINGLRRVLDELTLCRTDQELQYESLSEEMTYLKKNHEEEMKALQCAAGGNVNVEMNAAPGVDLAVLLNNMRAEYEALAEQNRKDAEAWFNEKSASLQQQISHDSGAATFARSQLTEMRRTLQTLEIQLQSLMATKHSLECSLTETESNYCTQLAQIQAQIGALEEQLHQVRTETEGQKLEYEHLLDVKVHLEKEIETYCRLIDGDGNSCSKSKGFGSGSPGNSSKDLSKTTLVKTVVEELDQRGKVLSSRIHSIEEKTSKMTNGKTEQRVPF
RUO
KRT28
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers