missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KLHL2 Partial ORF (NP_009177.2, 1 a.a. - 66 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011275-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194016

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

Sequence: METPPLPPACTKQGHQKPLDSKDDNTEKHCPVTVNPWHMKKAFKVMNELRSQNLLCDVTIVAEDME
Specifications

Specifications

NP_009177.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
METPPLPPACTKQGHQKPLDSKDDNTEKHCPVTVNPWHMKKAFKVMNELRSQNLLCDVTIVAEDME
RUO
KLHL2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
11275
KLHL2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ABP-KELCH/MAV/MAYVEN
KLHL2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers