missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KLF9 Partial ORF (NP_001197, 25 a.a. - 101 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000687-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195344

  • 349.00€ / 10µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a transcription factor that binds to GC box elements located in the promoter. Binding of the encoded protein to a single GC box inhibits mRNA expression while binding to tandemly repeated GC box elements activates transcription. [provided by RefSeq]

Sequence: PEHGVAPDAERLRLPEREVTKEHGDPGDTWKDYCTLVTIAKSLLDLNKYRPIQTPSVCSDSLESPDEDMGSDSDVTT
Specifications

Specifications

NP_001197
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.21kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PEHGVAPDAERLRLPEREVTKEHGDPGDTWKDYCTLVTIAKSLLDLNKYRPIQTPSVCSDSLESPDEDMGSDSDVTT
RUO
KLF9
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
687
KLF9 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BTEB/BTEB1
KLF9
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers