missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KLC2 Full-length ORF (NP_073733.1, 1 a.a. - 622 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00064837-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193763

  • 528.00€ / 25µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

Kinesin is a molecular motor that generates ATP-dependent movement of vesicles and organelles along microtubules. Kinesin consists of 2 light chains, such as KLC2, and 2 heavy chains (see KIF5B; MIM 602809) in a 1:1 stoichiometric ratio (Rahman et al., 1998 [PubMed 9624122]).[supplied by OMIM]

Sequence: MAMMVFPREEKLSQDEIVLGTKAVIQGLETLRGEHRALLAPLVAPEAGEAEPGSQERCILLRRSLEAIELGLGEAQVILALSSHLGAVESEKQKLRAQVRRLVQENQWLREELAGTQQKLQRSEQAVAQLEEEKQHLLFMSQIRKLDEDASPNEEKGDVPKDTLDDLFPNEDEQSPAPSPGGGDVSGQHGGYEIPARLRTLHNLVIQYASQGRYEVAVPLCKQALEDLEKTSGHDHPDVATMLNILALVYRDQNKYKEAAHLLNDALAIREKTLGKDHPAVAATLNNLAVLYGKRGKYKEAEPLCKRALEIREKVLGKFHPDVAKQLSNLALLCQNQGKAEEVEYYYRRALEIYATRLGPDDPNVAKTKNNLASCYLKQGKYQDAETLYKEILTRAHEKEFGSVNGDNKPIWMHAEEREESKDKRRDSAPYGEYGSWYKACKVDSPTVNTTLRSLGALYRRQGKLEAAHTLEDCASRNRKQGLDPASQTKVVELLKDGSGRRGDRRSSRDMAGGAGPRSESDLEDVGPTAEWNGDGSGSLRRSGSFGKLRDALRRSSEMLVKKLQGGTPQEPPNPRMKRASSLNFLNKSVEEPTQPGGTGLSDSRTLSSSSMDLSRRSSLVG
Specifications

Specifications

NP_073733.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
95.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ12387
KLC2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
64837
KLC2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAMMVFPREEKLSQDEIVLGTKAVIQGLETLRGEHRALLAPLVAPEAGEAEPGSQERCILLRRSLEAIELGLGEAQVILALSSHLGAVESEKQKLRAQVRRLVQENQWLREELAGTQQKLQRSEQAVAQLEEEKQHLLFMSQIRKLDEDASPNEEKGDVPKDTLDDLFPNEDEQSPAPSPGGGDVSGQHGGYEIPARLRTLHNLVIQYASQGRYEVAVPLCKQALEDLEKTSGHDHPDVATMLNILALVYRDQNKYKEAAHLLNDALAIREKTLGKDHPAVAATLNNLAVLYGKRGKYKEAEPLCKRALEIREKVLGKFHPDVAKQLSNLALLCQNQGKAEEVEYYYRRALEIYATRLGPDDPNVAKTKNNLASCYLKQGKYQDAETLYKEILTRAHEKEFGSVNGDNKPIWMHAEEREESKDKRRDSAPYGEYGSWYKACKVDSPTVNTTLRSLGALYRRQGKLEAAHTLEDCASRNRKQGLDPASQTKVVELLKDGSGRRGDRRSSRDMAGGAGPRSESDLEDVGPTAEWNGDGSGSLRRSGSFGKLRDALRRSSEMLVKKLQGGTPQEPPNPRMKRASSLNFLNKSVEEPTQPGGTGLSDSRTLSSSSMDLSRRSSLVG
RUO
KLC2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers