missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIAA1542 Partial ORF (NP_065952, 92 a.a. - 189 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057661-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192873

  • 528.00€ / 25µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

Sequence: KLEAAGSFNSDDDAESCPICLNAFRDQAVGTPENCAHYFCLDCIVEWSKNANSCPVDRTLFKCICIRAQFGGKILKKIPVENTKASEEEEDPTFCEVC
Specifications

Specifications

NP_065952
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KLEAAGSFNSDDDAESCPICLNAFRDQAVGTPENCAHYFCLDCIVEWSKNANSCPVDRTLFKCICIRAQFGGKILKKIPVENTKASEEEEDPTFCEVC
RUO
PHRF1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
57661
KIAA1542 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1542/RNF221
PHRF1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers