missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIAA1109 Full-length ORF (AAH45783.1, 1 a.a. - 191 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084162-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125673

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MQLDFKSIHVQEPQEPSLQDASLKPKVECSVVTEFTDHICVTMDAELIMFLHDLVSAYLKEKEKAIFPPRILSTRPGQKSPIIIHDDNSSDKDREDSITYTTVDWRDFMCNTWHLEPTLRLISWTGRKIDPVGVDYILQKLGFHHARTTIPKWLQRGVMDPLDKVLSVLIKKLGTALQDEKEKKGKDKEEH
Specifications

Specifications

AAH45783.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp781P0474/FSA/MGC110967
KIAA1109
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84162
KIAA1109 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQLDFKSIHVQEPQEPSLQDASLKPKVECSVVTEFTDHICVTMDAELIMFLHDLVSAYLKEKEKAIFPPRILSTRPGQKSPIIIHDDNSSDKDREDSITYTTVDWRDFMCNTWHLEPTLRLISWTGRKIDPVGVDYILQKLGFHHARTTIPKWLQRGVMDPLDKVLSVLIKKLGTALQDEKEKKGKDKEEH
RUO
KIAA1109
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers