missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIAA0971 Partial ORF (NP_055744, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00022868-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16104206

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein that is localized in the mitochondrial inner compartment and that may play a role in mitochondrial apoptosis. Nonsense mutations have been reported to result in cytochrome c oxidase deficiency. [provided by RefSeq]

Sequence: MLTTLKPFGSVSVESKMNNKAGSFFWNLRQFSTLVSTSRTMRLCCLGLCKPKIVHSNWNILNNFHNRMQSTDIIRYLFQDAFIFKSDVGFQTKGISTLTA
Specifications

Specifications

NP_055744
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MLTTLKPFGSVSVESKMNNKAGSFFWNLRQFSTLVSTSRTMRLCCLGLCKPKIVHSNWNILNNFHNRMQSTDIIRYLFQDAFIFKSDVGFQTKGISTLTA
RUO
FASTKD2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
22868
KIAA0971 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0971
FASTKD2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers