missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIAA0247 Full-length ORF (NP_055549.1, 1 a.a. - 303 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009766-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194082

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MCHGRIAPKSTSVFAVASVGHGVFLPLVILCTLLGDGLASVCPLPPEPENGGYICHPRPCRDPLTAGSVIEYLCAEGYMLKGDYKYLTCKNGEWKPAMEISCRLNEDKDTHTSLGVPTLSIVASTASSVALILLLVVLFVLLQPKLKSFHHSRRDQGVSGDQVSIMVDGVQVALPSYEEAVYGSSGHCVPPADPRVQIVLSEGSGPSGRSVPREQQLPDQGACSSAGGEDEAPGQSGLCEAWGSRASETVMVHQATTSSWVAGSGNRQLAHKETADSENSDIQSLLSLTSEEYTDDIPLLKEA
Specifications

Specifications

NP_055549.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
58.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0247
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
9766
KIAA0247 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MCHGRIAPKSTSVFAVASVGHGVFLPLVILCTLLGDGLASVCPLPPEPENGGYICHPRPCRDPLTAGSVIEYLCAEGYMLKGDYKYLTCKNGEWKPAMEISCRLNEDKDTHTSLGVPTLSIVASTASSVALILLLVVLFVLLQPKLKSFHHSRRDQGVSGDQVSIMVDGVQVALPSYEEAVYGSSGHCVPPADPRVQIVLSEGSGPSGRSVPREQQLPDQGACSSAGGEDEAPGQSGLCEAWGSRASETVMVHQATTSSWVAGSGNRQLAHKETADSENSDIQSLLSLTSEEYTDDIPLLKEA
RUO
KIAA0247
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers