missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KCTD13 Partial ORF (NP_849194, 24 a.a. - 119 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00253980-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: LEPGPAAYGLKPLTPNSKYVKLNVGGSLHYTTLRTLTGQDTMLKAMFSGRVEVLTDAGGWVLIDRSGRHFGTILNYLRDGSVPLPESTRELGELLGSpecifications
NP_849194 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.3kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LEPGPAAYGLKPLTPNSKYVKLNVGGSLHYTTLRTLTGQDTMLKAMFSGRVEVLTDAGGWVLIDRSGRHFGTILNYLRDGSVPLPESTRELGELLG | |
RUO | |
KCTD13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
253980 | |
KCTD13 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKSG86/PDIP1/POLDIP1 | |
KCTD13 | |
Recombinant | |
wheat germ expression system |