Learn More
Abnova™ Human KCNA1 Full-length ORF (NP_000208.2, 1 a.a. - 495 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003736-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a voltage-gated delayed potassium channel that is phylogenetically related to the Drosophila Shaker channel. The encoded protein has six putative transmembrane segments (S1-S6), and the loop between S5 and S6 forms the pore and contains the conserved selectivity filter motif (GYGD). The functional channel is a homotetramer. The N-terminus of the channel is associated with beta subunits that can modify the inactivation properties of the channel as well as affect expression levels. The C-terminus of the channel is complexed to a PDZ domain protein that is responsible for channel targeting. Mutations in this gene have been associated with myokymia with periodic ataxia (AEMK). [provided by RefSeq]
Sequence: MTVMSGENVDEASAAPGHPQDGSYPRQADHDDHECCERVVINISGLRFETQLKTLAQFPNTLLGNPKKRMRYFDPLRNEYFFDRNRPSFDAILYYYQSGGRLRRPVNVPLDMFSEEIKFYELGEEAMEKFREDEGFIKEEERPLPEKEYQRQVWLLFEYPESSGPARVIAIVSVMVILISIVIFCLETLPELKDDKDFTGTVHRIDNTTVIYNSNIFTDPFFIVETLCIIWFSFELVVRFFACPSKTDFFKNIMNFIDIVAIIPYFITLGTEIAEQEGNQKGEQATSLAILRVIRLVRVFRIFKLSRHSKGLQILGQTLKASMRELGLLIFFLFIGVILFSSAVYFAEAEEAESHFSSIPDAFWWAVVSMTTVGYGDMYPVTIGGKIVGSLCAIAGVLTIALPVPVIVSNFNYFYHRETEGEEQAQLLHVSSPNLASDSDLSRRSSSTMSKSEYMEIEEDMNNSIAHYRQVNIRTANCTTANQNCVNKSKLLTDVSpecifications
NP_000208.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
82.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AEMK/EA1/HBK1/HUK1/KV1.1/MBK1/MGC126782/MGC138385/MK1/RBK1 | |
KCNA1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3736 | |
KCNA1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTVMSGENVDEASAAPGHPQDGSYPRQADHDDHECCERVVINISGLRFETQLKTLAQFPNTLLGNPKKRMRYFDPLRNEYFFDRNRPSFDAILYYYQSGGRLRRPVNVPLDMFSEEIKFYELGEEAMEKFREDEGFIKEEERPLPEKEYQRQVWLLFEYPESSGPARVIAIVSVMVILISIVIFCLETLPELKDDKDFTGTVHRIDNTTVIYNSNIFTDPFFIVETLCIIWFSFELVVRFFACPSKTDFFKNIMNFIDIVAIIPYFITLGTEIAEQEGNQKGEQATSLAILRVIRLVRVFRIFKLSRHSKGLQILGQTLKASMRELGLLIFFLFIGVILFSSAVYFAEAEEAESHFSSIPDAFWWAVVSMTTVGYGDMYPVTIGGKIVGSLCAIAGVLTIALPVPVIVSNFNYFYHRETEGEEQAQLLHVSSPNLASDSDLSRRSSSTMSKSEYMEIEEDMNNSIAHYRQVNIRTANCTTANQNCVNKSKLLTDV | |
RUO | |
KCNA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |