Learn More
R&D Systems™ Human KAT2A/GCN5 (NP_066564.2) VersaClone cDNA
Specifications
Specifications
| Gene | KAT2A/GCN5 |
| Accession Number | NP_066564.2 |
| Gene ID (Entrez) | 2648 |
| Gene Alias | EC 2.3.1.48, GCN5 general control of amino-acid synthesis 5-like 2 (yeast), GCN5L2, GCN5L2GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2, GCN5MGC102791, General control of amino acid synthesis protein 5-like 2, General control of amino acid synthesis, yeast, homolog-like 2, hGCN5, Histone acetyltransferase GCN5, histone acetyltransferase KAT2A, hsGCN5, K(lysine) acetyltransferase 2 A, KAT2A, Lysine acetyltransferase 2 A, PCAF-b, STAF97 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |