missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Kallikrein 1 ELISA Kit (Colorimetric)

Product Code. 18699600 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18699600

Brand: Novus Biologicals™ KA2009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Assay Range 156 to 10000 pg/mL
Assay Sensitivity <10 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
For Use With (Application) ELISA, Quantification
Gene ID (Entrez) 3816
Gene Symbol KLK1
Quantity 1 Kit
Research Discipline Cancer
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Synonym EC 3.4.21, EC 3.4.21.35, glandular kallikrein 1, hK1, kallikrein 1, kallikrein 1, renal/pancreas/salivary, kallikrein serine protease 1, kallikrein-1, Kidney/pancreas/salivary gland kallikrein, Klk6, KLKR, Tissue kallikrein
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.