missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ITGA4 Partial ORF (NP_000876, 98 a.a. - 207 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003676-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16117001

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

The product of this gene belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes an alpha 4 chain. Unlike other integrin alpha chains, alpha 4 neither contains an I-domain, nor undergoes disulfide-linked cleavage. Alpha 4 chain associates with either beta 1 chain or beta 7 chain. [provided by RefSeq]

Sequence: RIGKNPGQTCEQLQLGSPNGEPCGKTCLEERDNQWLGVTLSRQPGENGSIVTCGHRWKNIFYIKNENKLPTGGCYGVPPDLRTELSKRIAPCYQDYVKKFGENFASCQAG
Specifications

Specifications

NP_000876
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RIGKNPGQTCEQLQLGSPNGEPCGKTCLEERDNQWLGVTLSRQPGENGSIVTCGHRWKNIFYIKNENKLPTGGCYGVPPDLRTELSKRIAPCYQDYVKKFGENFASCQAG
RUO
ITGA4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3676
ITGA4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD49D/IA4/MGC90518
ITGA4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers