missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ITGA2 Partial ORF (NP_002194.2, 30 a.a. - 119 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003673-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16156991

  • 335.00€ / 10µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

This gene product belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane glycoproteins composed of a distinct alpha chain and a common beta chain. They are found on a wide variety of cell types including, T cells, fibroblasts and platelets. Integrins are involved in cell adhesion and also participate in cell-surface mediated signalling. [provided by RefSeq]

Sequence: YNVGLPEAKIFSGPSSEQFGYAVQQFINPKGNWLLVGSPWSGFPENRMGDVYKCPVDLSTATCEKLNLQTSTSIPNVTEMKTNMSLGLIL
Specifications

Specifications

NP_002194.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.53kDa
12.5% SDS-PAGE Stained with Coomassie Blue
YNVGLPEAKIFSGPSSEQFGYAVQQFINPKGNWLLVGSPWSGFPENRMGDVYKCPVDLSTATCEKLNLQTSTSIPNVTEMKTNMSLGLIL
RUO
ITGA2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3673
ITGA2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BR/CD49B/GPIa/VLA-2/VLAA2
ITGA2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers