missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IQGAP2 Partial ORF (NP_006624, 343 a.a. - 449 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010788-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105712

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the IQGAP family. The protein contains three IQ domains, one calponin homology domain, one Ras-GAP domain and one WW domain. It interacts with components of the cytoskeleton, with cell adhesion molecules, and with several signaling molecules to regulate cell morphology and motility. [provided by RefSeq]

Sequence: MYQNELFNLQKQNTMNYLAHEELLIAVEMLSAVALLNQALESNDLVSVQNQLRSPAIGLNNLDKAYVERYANTLLSVKLEVLSQGQDNLSWNEIQNCIDMVNAQIQE
Specifications

Specifications

NP_006624
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.51kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MYQNELFNLQKQNTMNYLAHEELLIAVEMLSAVALLNQALESNDLVSVQNQLRSPAIGLNNLDKAYVERYANTLLSVKLEVLSQGQDNLSWNEIQNCIDMVNAQIQE
RUO
IQGAP2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
10788
IQGAP2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IQGAP2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers