Learn More
Abnova™ Human INPP5B Full-length ORF (AAH58932.1, 1 a.a. - 748 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003633-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Cellular calcium signaling is controlled by the production of inositol phosphates (IPs) by phospholipase C in response to extracellular signals. The IP signaling molecules are inactivated by a family of inositol polyphosphate-5-phosphatases (5-phosphatases). This gene encodes the type II 5-phosphatase. The protein is localized to the cytosol and mitochondria, and associates with membranes through an isoprenyl modification near the C-terminus. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: MDQSVAIQETLAEGEYCVIAVQGVLCEGDSRQSRLLGLVRYRLEHGGQEHALFLYTHRRMAITGDDVSLDQIVPVSRDFTLEEVSPDGELYILGSDVTVQLDTAELSLVFQLPFGSQTRMFLHEVARACPGFDSATRDPEFLWLSRYRCAELELEMPTPRGCNSALVTWPGYATIGGGGSNFDGLRPNGKGVPMDQSSRGQDKPESLQPRQNKSKSEITDMVRSSTITVSDKAHILSMQKFGLRDTIVKSHLLQKEEDYTYIQNFRFFAGTYNVNGQSPKECLRLWLSNGIQAPDVYCVGFQELDLSKEAFFFHDTPKEEEWFKAVSEGLHPDAKYAKVKLIRLVGIMLLLYVKQEHAAYISEVEAETVGTGIMGRMGNKGGVAIRFQFHNTSICVVNSHLAAHIEEYERRNQDYKDICSRMQFCQPDPSLPPLTISNHDVILWLGDLNYRIEELDVEKVKKLIEEKDFQMLYAYDQLKIQVAAKTVFEGFTEGELTFQPTYKYDTGSDDWDTSEKCRAPAWCDRILWKGKNITQLSYQSHMALKTSDHKPVSSVFDIGVRVVNDELYRKTLEEIVRSLDKMENANIPSVSLSKREFCFQNVKYMQLKVESFTIHNGQVPCHFEFINKPDEESYCKQWLNANPSRGFLLPDSDVEIDLELFVNKTTATKLNSGEDKIEDILVLHLDRGKDYFLSVSGNYLPSCFGSPIHTLCYMREPILDLPLETISELLAYLAAYCFETQLVTKSLISpecifications
AAH58932.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
111.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
5PTase/MGC65156/MGC71303 | |
INPP5B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3633 | |
INPP5B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDQSVAIQETLAEGEYCVIAVQGVLCEGDSRQSRLLGLVRYRLEHGGQEHALFLYTHRRMAITGDDVSLDQIVPVSRDFTLEEVSPDGELYILGSDVTVQLDTAELSLVFQLPFGSQTRMFLHEVARACPGFDSATRDPEFLWLSRYRCAELELEMPTPRGCNSALVTWPGYATIGGGGSNFDGLRPNGKGVPMDQSSRGQDKPESLQPRQNKSKSEITDMVRSSTITVSDKAHILSMQKFGLRDTIVKSHLLQKEEDYTYIQNFRFFAGTYNVNGQSPKECLRLWLSNGIQAPDVYCVGFQELDLSKEAFFFHDTPKEEEWFKAVSEGLHPDAKYAKVKLIRLVGIMLLLYVKQEHAAYISEVEAETVGTGIMGRMGNKGGVAIRFQFHNTSICVVNSHLAAHIEEYERRNQDYKDICSRMQFCQPDPSLPPLTISNHDVILWLGDLNYRIEELDVEKVKKLIEEKDFQMLYAYDQLKIQVAAKTVFEGFTEGELTFQPTYKYDTGSDDWDTSEKCRAPAWCDRILWKGKNITQLSYQSHMALKTSDHKPVSSVFDIGVRVVNDELYRKTLEEIVRSLDKMENANIPSVSLSKREFCFQNVKYMQLKVESFTIHNGQVPCHFEFINKPDEESYCKQWLNANPSRGFLLPDSDVEIDLELFVNKTTATKLNSGEDKIEDILVLHLDRGKDYFLSVSGNYLPSCFGSPIHTLCYMREPILDLPLETISELLAYLAAYCFETQLVTKSLI | |
RUO | |
INPP5B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |