missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human INHA Partial ORF (AAH06391, 234 a.a. - 333 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003623-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100175

  • 508.00€ / 25µg
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

The inhibin alpha subunit joins either the beta A or beta B subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumour-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. However, in prostate cancer, expression of the inhibin alpha-subunit gene was suppressed and was not detectable in poorly differentiated tumor cells. Furthermore, because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone. [provided by RefSeq]

Sequence: TPLMSWPWSPSALRLLQRPPEEPAAHANCHRVALNISFQELGWERWIVYPPSFIFHYCHGGCGLHIPPNLSLPVPGAPPTPAQPYSLLPGAQPCCAALPG
Specifications

Specifications

AAH06391
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TPLMSWPWSPSALRLLQRPPEEPAAHANCHRVALNISFQELGWERWIVYPPSFIFHYCHGGCGLHIPPNLSLPVPGAPPTPAQPYSLLPGAQPCCAALPG
RUO
INHA
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
3623
INHA (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
INHA
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers