Learn More
Abnova™ Human ING5 Full-length ORF (NP_115705.2, 1 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084289-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This protein can bind TP53 and EP300/p300, a component of the histone acetyl transferase complex, suggesting its involvement in TP53-dependent regulatory pathway. [provided by RefSeq]
Sequence: MATAMYLEHYLDSIENLPCELQRNFQLMRELDQRTEDKKAEIDILAAEYISTVKTLSPDQRVERLQKIQNAYSKCKEYSDDKVQLAMQTYEMVDKHIRRLDADLARFEADLKDKMEGSDFESSGGRGLKKGRGQKEKRGSRGRGRRTSEEDTPKKKKHKGGSEFTDTILSVHPSDVLDMPVDPNEPTYCLCHQVSYGEMIGCDNPDCPIEWFHFACVDLTTKPKGKWFCPRCVQEKRKKKSpecifications
NP_115705.2 | |
Liquid | |
84289 | |
ING5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATAMYLEHYLDSIENLPCELQRNFQLMRELDQRTEDKKAEIDILAAEYISTVKTLSPDQRVERLQKIQNAYSKCKEYSDDKVQLAMQTYEMVDKHIRRLDADLARFEADLKDKMEGSDFESSGGRGLKKGRGQKEKRGSRGRGRRTSEEDTPKKKKHKGGSEFTDTILSVHPSDVLDMPVDPNEPTYCLCHQVSYGEMIGCDNPDCPIEWFHFACVDLTTKPKGKWFCPRCVQEKRKKK | |
RUO | |
ING5 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23842/p28ING5 | |
ING5 | |
Yes | |
wheat germ expression system |