missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ING4 Full-length ORF (AAH07781.1, 1 a.a. - 249 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH07781.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51147 |
Molecular Weight (g/mol) | 54.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16159692
|
Abnova™
H00051147-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16149692
|
Abnova™
H00051147-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a tumor suppressor protein that contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This protein can bind TP53 and EP300/p300, a component of the histone acetyl transferase complex, suggesting its involvement in the TP53-dependent regulatory pathway. Multiple alternatively spliced transcript variants have been observed that encode distinct proteins. [provided by RefSeq]
Sequence: MAAGMYLEHYLDSIENLPFELQRNFQLMRDLDQRTEDLKAEIDKLATEYMSSARSLSSEEKLALLKQIQEAYGKCKEFGDDKVQLAMQTYEMVDKHIRRLDTDLARFEADLKEKQIESSDYDSSSSKGKKKGRTQKEKKAARARSKGKNSDEEAPKTAQKKLKLVRTSPEYGMPSVTFGSVHPSDVLDMPVDPNEPTYCLCHQVSYGEMIGCDNPDCSIEWFHFACVGLTTKPRGKWFCPRCSQERKKKSpecifications
AAH07781.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAAGMYLEHYLDSIENLPFELQRNFQLMRDLDQRTEDLKAEIDKLATEYMSSARSLSSEEKLALLKQIQEAYGKCKEFGDDKVQLAMQTYEMVDKHIRRLDTDLARFEADLKEKQIESSDYDSSSSKGKKKGRTQKEKKAARARSKGKNSDEEAPKTAQKKLKLVRTSPEYGMPSVTFGSVHPSDVLDMPVDPNEPTYCLCHQVSYGEMIGCDNPDCSIEWFHFACVGLTTKPRGKWFCPRCSQERKKK | |
RUO | |
ING4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51147 | |
ING4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC12557/my036/p29ING4 | |
ING4 | |
Recombinant | |
wheat germ expression system |