Learn More
Abnova™ Human IMMP1L Full-length ORF (NP_659418.1, 1 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00196294-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The mitochondrial inner membrane peptidase (IMP) complex generates mature, active proteins in the mitochondrial intermembrane space by proteolytically removing the mitochondrial targeting presequence of nuclear-encoded proteins. IMP1 and IMP2 (IMMP2L; MIM 605977) are the catalytic subunits of the IMP complex (Burri et al., 2005 [PubMed 15814844]).[supplied by OMIM]
Sequence: MLRGVLGKTFRLVGYTIQYGCIAHCAFEYVGGVVMCSGPSMEPTIQNSDIVFAENLSRHFYGIQRGDIVIAKSPSDPKSNICKRVIGLEGDKILTTSPSDFFKSHSYVPMGHVWLEGDNLQNSTDSRCYGPIPYGLIRGRIFFKIWPLSDFGFLRASPNGHRFSDDSpecifications
NP_659418.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ25059/IMP1/IMP1-LIKE | |
IMMP1L | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
196294 | |
IMMP1L (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLRGVLGKTFRLVGYTIQYGCIAHCAFEYVGGVVMCSGPSMEPTIQNSDIVFAENLSRHFYGIQRGDIVIAKSPSDPKSNICKRVIGLEGDKILTTSPSDFFKSHSYVPMGHVWLEGDNLQNSTDSRCYGPIPYGLIRGRIFFKIWPLSDFGFLRASPNGHRFSDD | |
RUO | |
IMMP1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |