missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IL17E Full-length ORF (NP_073626.1, 1 a.a. - 177 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_073626.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 64806 |
Molecular Weight (g/mol) | 46.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16123763
|
Abnova™
H00064806-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment:
Log in to see stock. |
|||||
16113763
|
Abnova™
H00064806-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment:
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. This cytokine can induce NF-kappaB activation, and stimulate the production of IL8. Both this cytokine and IL17B are ligands for the cytokine receptor IL17BR. Studies of the similar gene in mice suggested that this cytokine may be a proinflammatory cytokine favoring Th2-type immune response. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq]
Sequence: MRERPRLGEDSSLISLFLQVVAFLAMVMGTHTYSHWPSCCPSKGQDTSEELLRWSTVPVPPLEPARPNRHPESCRASEDGPLNSRAISPWRYELDRDLNRLPQDLYHARCLCPHCVSLQTGSHMDPRGNSELLYHNQTVFYRRPCHGEKGTHKGYCLERRLYRVSLACVCVRPRVMGSpecifications
NP_073626.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
46.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MRERPRLGEDSSLISLFLQVVAFLAMVMGTHTYSHWPSCCPSKGQDTSEELLRWSTVPVPPLEPARPNRHPESCRASEDGPLNSRAISPWRYELDRDLNRLPQDLYHARCLCPHCVSLQTGSHMDPRGNSELLYHNQTVFYRRPCHGEKGTHKGYCLERRLYRVSLACVCVRPRVMG | |
RUO | |
IL25 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
64806 | |
IL17E (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL-17E/IL-25/IL17E | |
IL25 | |
Recombinant | |
wheat germ expression system |