missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ anti-Interleukin 6 Recombinant

Product Code. 10117493
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10117493

Brand: Thermo Scientific™ SIL6

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Interleukin 6 Recombinant protein SIL6 from Thermo Scientific™ specifically detects Interleukin 6 in Not Applicable samples, and it is validated for Control, ELISA and WB

Caveolae are specialized domains of the plasma membrane that are implicated in the sequestration of a variety of lipid and protein molecules. It has been suggested that these important cellular organelles have a pivotal role in such diverse biochemical processes as lipid metabolism, growth regulation, signal transduction, and apoptosis. Caveolin interacts with and regulates heterotrimeric G-proteins. Currently, there are three members of the caveolin multigene family which are known to encode 21-24 kDa integral membrane proteins that comprise the major structural component of the caveolar membrane in vivo. Caveolin-2 protein is abundantly expressed in fibroblasts and differentiated adipocytes, smooth and skeletal muscle, and endothelial cells. The expression of caveolin-1 is similar to that of caveolin-2 while caveolin-3 expression appears to be limited to muscle tissue types.
Host Species: E. Coli
Cross Reactivity: Not Applicable
Formulation: 1 Vial purified Protein (1 ng/ml) in 4% BSA and no preservative;

PA5-32691 detects CAV1/Caveolin in Human samples. PA5-32691 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-32691 The immunogen is Synthetic 16 amino acid peptide from N-terminus of human CAV1/Caveolin..
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 10.2 to 400 pg/mL
Assay Sensitivity <1 pg/mL
Description Human IL-6 Standard for ELISA
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine
Specificity SIL6
Sufficient For ELISA standard curve with replicates
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
Quantity 1 Vial
Sample Volume 50 μL
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Target Natural and recombinant human interleukin-6 (IL-6)
Total Assay Time 3 hr.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.