missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-36 alpha/IL-1F6 DuoSet ELISA

Product Code. 30020844
Click to view available options
Format:
96-well
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30020844

Brand: R&D Systems™ DY107805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For determination of natural and recombinant Interleukin 36 alpha (IL-36α) in humans.

The Human IL-36 alpha/IL-1F6 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-36 alpha/IL-1F6.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Solid Phase Sandwich ELISA
Species Human
Binding Target IL-36 alpha/IL-1F6
Assay Range 12.50 to 800 pg/mL
Format 96-well
Gene ID (Entrez) 27179 (Human); 54448 (Mouse)
Kit Contents 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.