missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-36 alpha/IL-1F6 DuoSet ELISA

Product Code. 18750498
Change view
Click to view available options
Format:
96-well
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
18750498 96-well Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18750498 Supplier R&D Systems™ Supplier No. DY107805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For determination of natural and recombinant Interleukin 36 alpha (IL-36α) in humans.

The Human IL-36 alpha/IL-1F6 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-36 alpha/IL-1F6.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Solid Phase Sandwich ELISA
Species Human
Binding Target IL-36 alpha/IL-1F6
Assay Range 12.50 to 800 pg/mL
Format 96-well
Gene ID (Entrez) 27179 (Human); 54448 (Mouse)
Kit Contents 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.