Learn More
ACROBiosystems Human IL-2 R alpha / CD25 Protein, Fc Tag
HPLC-verified
Brand: ACROBiosystems ILA-H5251-100UG
Additional Details : Weight : 0.00001kg
Description
Interleukin-2 receptor subunit alpha (IL2RA) is also known as IL-2R subunit alpha, IL-2-RA, IL2-RA, TAC antigen, p55, CD antigen CD25, is a type I transmembrane glycoprotein. IL2RA is expressed on activated T cells and regulatory T cells, and is capable of binding IL2 with low affinity by itself. However, a ligand-induced high affinity heterotrimeric receptor complex is produced when IL2RA is associated non-covelently with the IL2 receptor beta and gamma chain, and subsequently initiates the intacellular signal pathways such as MAPK or JAK/STAT. On dendritic cells (DC), CD25 has been previously regarded as an activation marker, while both murine and human DC can express CD25, they do not express the beta-chain of the IL-2 receptor, which is indispensable for the execution of IL-2 signaling.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.4 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_000408 | |
100 μg | |
Glu 22 - Cys 213 | |
IL2RA,CD25,p55,IL2-RA,IL-2-RA | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
48.4 kDa | |
Human IL-2 R alpha Protein, Fc Tag (HPLC-verified) (ILA-H5251) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession # NP_000408). | |
-20°C | |
IL-2 R alpha | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |