Learn More
R&D Systems™ Human IL-17 Quantikine HS ELISA Kit
Product Code. 18730877
Change view
Click to view available options
Content And Storage:
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
18730877 Store the unopened product at 2°C to 8°C. Do not use past expiration date. Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18730877 Supplier R&D Systems™ Supplier No. HS170
  • ONLINE EXCLUSIVE PRICE
    1109.64€
    991.20€ / Pack of 96
    Save 118.44€
    10% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Human IL-17 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human IL-17 Quantikine HS ELISA Kit quantifies human IL-17/IL-17A in serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Type Human IL-17 Quantikine HS ELISA Kit
Gene ID (Entrez) 3605 (Human); 16171 (Mouse); 301289 (Rat); 481837 (Canine)
Includes Microplate, conjugate, standards, assay diluents, calibrator diluents, wash buffer, color reagent, stop Solution, plate sealers
Intended Use Measure IL-17 levels in serum and plasma
Product Line R&D Systems
Time Range 4 hours