Learn More
R&D Systems™ Human IL-15, Carrier-Free Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 247-ILB-025/CF
Description
Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. The ED50 for this effect is 0.3-2.6 ng/mL. The specific activity of Recombinant Human IL-15 is approximately 4.5 x 105 U/μg, which is calibrated against recombinant human IL-15 WHO International Standard (NIBSC code: 95/554). Specific activity is for reference purposes only and is not routinely tested.Specifications
| P40933.1 | |
| 3600 | |
| 25μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| IL15, IL-15MGC9721, interleukin 15, interleukin-15 | |
| IL15 | |
| Unconjugated |
| Lyophilized from a 0.2μm filtered solution in PBS. Reconstitute at 100μg/mL in sterile PBS. | |
| Observed M.W.: 10kDa, reducing conditions, Theoretical M.W.: 13kDa | |
| E. coli-derived human IL-15 protein Asn49-Ser162 | |
| <0.10 EU per 1μg of the protein by the LAL method. | |
| IL-15 | |
| Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. The ED50 for this effect is typically 0.3-2.6ng/mL. The specific activity of Recombinant Human IL-15 is approximately 4.5 x 105 U/μg, which is calibrated against recombinant human IL-15 WHO International Standard (NIBSC code: 95/554). | |
| Yes |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.