Learn More
R&D Systems™ Human IL-13 High Sensitivity Magnetic Luminex™ Performance Assay
Product Code. 18491610
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18491610 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18491610 Supplier R&D Systems™ Supplier No. LBHS213
Magnetic bead-based multiplex assay for the Luminex™ MAGPIX™ platform
  • Magnetic format allows for easier wash steps
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
  • Requires a small sample volume (<50 μL)
  • Run your assay in just 3-5 hours
  • Mass-calibrated standards for consistent results with every new lot of material
  • Enough reagents for one 96-well plate
TRUSTED_SUSTAINABILITY
Specifications
Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target IL-13
Bead Type Magnetic
Assay Sensitivity 2.01pg/mL
Assay Range 6.25 - 25600pg/mL (Serum, Heparin Plasma, EDTA Plasma), 6.25 - 6400pg/mL (Cell Culture Supernates)
Gene ID (Entrez) 3596
Gene Symbol IL13
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Sample Volume Cell Culture Supernates (100μL),Serum (50μL),EDTA Plasma (50μL),Heparin Plasma (50μL)
Kit Contents
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Content And Storage Store the unopened product at 2° to 8°C. Do not use past expiration date.